Piperonal, 99%, Thermo Scientific Chemicals
Piperonal, 99%, Thermo Scientific Chemicals
Piperonal, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ACR131341000
Price (MYR)
87.69
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
87.69
Add to cart
Specifications
Chemical Name or MaterialPiperonal
CAS120-57-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement May cause an allergic skin reaction.
Health Hazard 3GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.