Phthalic acid, 99%, Thermo Scientific Chemicals
Phthalic acid, 99%, Thermo Scientific Chemicals
Phthalic acid, 99%, Thermo Scientific Chemicals
Phthalic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phthalic acid, 99%, Thermo Scientific Chemicals
CAS: 88-99-3 | C8H6O4 | 166.132 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
250 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR131070025
Price (MYR)
1,549.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,549.00
Add to cart
Chemical Identifiers
CAS141-86-6
IUPAC Namepyridine-2,6-diamine
Molecular FormulaC5H7N3
InChI KeyVHNQIURBCCNWDN-UHFFFAOYSA-N
SMILESNC1=CC=CC(N)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or flakes
Appearance (Color)Beige to grey to dark brown
AppearanceMay darken on storage
Infrared spectrumConforms
Melting point115°C to 123°C