Phthalic acid, 99%, Thermo Scientific Chemicals
Phthalic acid, 99%, Thermo Scientific Chemicals
Phthalic acid, 99%, Thermo Scientific Chemicals
Phthalic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phthalic acid, 99%, Thermo Scientific Chemicals
CAS: 88-99-3 | C8H6O4 | 166.132 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR1310700101 kgPlastic bottle
Catalog number ACR131070010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS6921-98-8
IUPAC NameN-(formamidomethyl)formamide
Molecular FormulaC3H6N2O2
InChI KeyQPJQPYQZFKFTHG-UHFFFAOYSA-N
SMILESO=CNCNC=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White
Assay (HPLC)≥96.0%
Melting Point (clear melt)139.0-145.0?C