Phthalhydrazide, 99%, Thermo Scientific Chemicals
Phthalhydrazide, 99%, Thermo Scientific Chemicals
Phthalhydrazide, 99%, Thermo Scientific Chemicals
Phthalhydrazide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phthalhydrazide, 99%, Thermo Scientific Chemicals
CAS: 1445-69-8 | C8H6N2O2 | 162.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number ACR131060250
Price (MYR)
475.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
475.00
Add to cart
Chemical Identifiers
CAS13759-92-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystals
Total conc. of imp.as REO=<0.1 %