Phloroglucinol Dihydrate, 99%, Thermo Scientific Chemicals
Phloroglucinol Dihydrate, 99%, Thermo Scientific Chemicals
Phloroglucinol Dihydrate, 99%, Thermo Scientific Chemicals
Phloroglucinol Dihydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phloroglucinol Dihydrate, 99%, Thermo Scientific Chemicals
CAS: 6099-90-7 | C6H6O3·2H2O | 162.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass Bottle
Catalog number ACR131041000
Price (MYR)
633.00
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
633.00
Add to cart
Chemical Identifiers
CAS92-31-9
IUPAC Name3-amino-7-(dimethylamino)-2-methyl-5λ⁴-phenothiazin-5-ylium chloride
Molecular FormulaC15H16ClN3S
InChI KeyHNONEKILPDHFOL-UHFFFAOYSA-M
SMILES[Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Dye)(as is basis): 65% min
Loss on Drying8.0% max
Solution Test(0.1% distilled water): Passes Test
Specific Absorbance (E 1%/1cm)(lambda max 650nm): >1200