Phenyl sulfide, 99%, Thermo Scientific Chemicals
Phenyl sulfide, 99%, Thermo Scientific Chemicals
Phenyl sulfide, 99%, Thermo Scientific Chemicals
Phenyl sulfide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyl sulfide, 99%, Thermo Scientific Chemicals
CAS: 139-66-2 | C12H10S | 186.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number ACR130951000
Price (MYR)
1,116.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,116.00
Add to cart
Chemical Identifiers
CAS554-12-1
IUPAC Namemethyl propanoate
Molecular FormulaC4H8O2
InChI KeyRJUFJBKOKNCXHH-UHFFFAOYSA-N
SMILESCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.3750 to 1.3790 (20°C, 589 nm)
Water=<0.3 % (K.F.)