Phenyl ether, 99%, Thermo Scientific Chemicals
Phenyl ether, 99%, Thermo Scientific Chemicals
Phenyl ether, 99%, Thermo Scientific Chemicals
Phenyl ether, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyl ether, 99%, Thermo Scientific Chemicals
CAS: 101-84-8 | C12H10O | 170.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
Catalog number ACR130601000
Price (MYR)
391.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
391.00
Add to cart
Chemical Identifiers
CAS1806-26-4
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)White
FormCrystals or powder or crystalline powder