Phenyl ether, 99%, Thermo Scientific Chemicals
Phenyl ether, 99%, Thermo Scientific Chemicals
Phenyl ether, 99%, Thermo Scientific Chemicals
Phenyl ether, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyl ether, 99%, Thermo Scientific Chemicals
CAS: 101-84-8 | C12H10O | 170.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR130600010
Price (MYR)
583.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
583.00
Add to cart
Chemical Identifiers
CAS58640-01-0
IUPAC Nametert-butyl 4-aminobutanoate hydrochloride
Molecular FormulaC8H18ClNO2
InChI KeyCZVNLCOJFFYZPG-UHFFFAOYSA-N
SMILESCl.CC(C)(C)OC(=O)CCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%