Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
CAS: 98-80-6 | C6H7BO2 | 121.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Packaging:
Glass bottle
Catalog number ACR130360500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS117-99-7
IUPAC Name2-benzoylphenol
Molecular FormulaC13H10O2
InChI KeyHJIAMFHSAAEUKR-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Identification (FTIR)Conforms
Melting Point (clear melt)36.0-42.0?C
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%