Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
CAS: 98-80-6 | C6H7BO2 | 121.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number ACR130360100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS9003-01-4
IUPAC NamePoly(acrylic acid)
Molecular Formula(C3H4O2)n
SMILESOC(=O)C(-*)C-*
MDL NumberMFCD00084394
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Assay48 to 52 % Solids
Molecular weight3000 to 5000 (approx)
Appearance (Form)Clear to hazy liquid