4-Phenoxyaniline, 97%, Thermo Scientific Chemicals
4-Phenoxyaniline, 97%, Thermo Scientific Chemicals
4-Phenoxyaniline, 97%, Thermo Scientific Chemicals
4-Phenoxyaniline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Phenoxyaniline, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number ACR130231000
Price (MYR)
687.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
687.00
Add to cart
Chemical Identifiers
CAS591-49-1
IUPAC Name1-methylcyclohex-1-ene
Molecular FormulaC7H12
InChI KeyCTMHWPIWNRWQEG-UHFFFAOYSA-N
SMILESCC1=CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.4492 to 1.4512 (20°C, 589 nm)
Stabilizer10 ppm hydroquinone