Phenoxazine, 97%, Thermo Scientific Chemicals
Phenoxazine, 97%, Thermo Scientific Chemicals
Phenoxazine, 97%, Thermo Scientific Chemicals
Phenoxazine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenoxazine, 97%, Thermo Scientific Chemicals
CAS: 135-67-1 | C12H9NO | 183.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR130220050
Price (MYR)
1,417.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,417.00
Add to cart
Chemical Identifiers
CAS98-58-8
IUPAC Name4-bromobenzene-1-sulfonyl chloride
Molecular FormulaC6H4BrClO2S
InChI KeyKMMHZIBWCXYAAH-UHFFFAOYSA-N
SMILESClS(=O)(=O)C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Melting point74°C to 77°C
Appearance (Color)White to beige
Infrared spectrumConforms
Titration Argentometric>=97.5 % (Ex CL)