Phenothiazine, 99%, Thermo Scientific Chemicals
Phenothiazine, 99%, Thermo Scientific Chemicals
Phenothiazine, 99%, Thermo Scientific Chemicals
Phenothiazine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenothiazine, 99%, Thermo Scientific Chemicals
CAS: 92-84-2 | C12H9NS | 199.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
500 g
Packaging:
View all
Plastic drum
Catalog number ACR130210050
Price (MYR)
2,215.00
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
2,215.00
Add to cart
Chemical Identifiers
CAS2423-71-4
IUPAC Name2,6-dimethyl-4-nitrophenol
Molecular FormulaC8H9NO3
InChI KeyFNORUNUDZNWQFF-UHFFFAOYSA-N
SMILESCC1=CC(=CC(C)=C1O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Orange to brown
Assay (GC)≥97.5%