1,10-Phenanthroline monohydrate, 99+%, pure, Thermo Scientific Chemicals
1,10-Phenanthroline monohydrate, 99+%, pure, Thermo Scientific Chemicals
1,10-Phenanthroline monohydrate, 99+%, pure, Thermo Scientific Chemicals
1,10-Phenanthroline monohydrate, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,10-Phenanthroline monohydrate, 99+%, pure, Thermo Scientific Chemicals
CAS: 5144-89-8 | C12H10N2O | 198.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR130135000
Price (MYR)
12,177.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
12,177.00
Add to cart
Chemical Identifiers
CAS156-57-0
IUPAC Namehydrogen 2-aminoethane-1-thiol chloride
Molecular FormulaC2H8ClNS
InChI KeyOGMADIBCHLQMIP-UHFFFAOYSA-N
SMILES[H+].[Cl-].NCCS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Loss on drying=<1 % (50°C, 4 h, 100 mmHg)
Appearance (Form)Crystalline powder or crystals and/or lumps
Melting point66°C to 70°C
Infrared spectrumConforms
View more
Indicator
RUO – Research Use Only