Phenanthrene, 97%, Thermo Scientific Chemicals
Phenanthrene, 97%, Thermo Scientific Chemicals
Phenanthrene, 97%, Thermo Scientific Chemicals
Phenanthrene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenanthrene, 97%, Thermo Scientific Chemicals
CAS: 85-01-8 | C14H10 | 178.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
500 g
Packaging:
View all
Glass bottle
Catalog number ACR130090050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS954-81-4
IUPAC Name2-(5-bromopentyl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC13H14BrNO2
InChI KeyQKVHAKICMNABGB-UHFFFAOYSA-N
SMILESBrCCCCCN1C(=O)C2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals
Appearance (Color)White to cream or pale yellow
Melting Point (clear melt)54-64?C
Assay (GC)≥96.0%