Pamoic acid, 98+%, Thermo Scientific Chemicals
Pamoic acid, 98+%, Thermo Scientific Chemicals
Pamoic acid, 98+%, Thermo Scientific Chemicals
Pamoic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pamoic acid, 98+%, Thermo Scientific Chemicals
CAS: 130-85-8 | C23H16O6 | 388.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number ACR129721000
Price (MYR)
570.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
570.00
Add to cart
Specifications
Chemical Name or MaterialPamoic acid
CAS130-85-8
Melting Point>300.0°C
Health Hazard 1Warning
Health Hazard 2GHS P Statement