Oxalic acid dihydrate, 99+%, extra pure, Thermo Scientific Chemicals
Oxalic acid dihydrate, 99+%, extra pure, Thermo Scientific Chemicals
Oxalic acid dihydrate, 99+%, extra pure, Thermo Scientific Chemicals
Oxalic acid dihydrate, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Oxalic acid dihydrate, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 6153-56-6 | C2H6O6 | 126.064 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 kg
Packaging:
View all
Plastic Bottle
Catalog number ACR129601000
Price (MYR)
365.00
Add to cart
Quantity:
100 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
365.00
Add to cart
Chemical Identifiers
CAS75-75-2
IUPAC Namemethanesulfonic acid
Molecular FormulaCH4O3S
InChI KeyAFVFQIVMOAPDHO-UHFFFAOYSA-N
SMILESCS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid or low melting solid
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0% (non-U.S. sourced material)
Assay from Supplier's CofA≥98.0% (U.S. sourced material)
CommentMaterial sourced in the U.S. and in other countries
Identification (FTIR)Conforms (non-U.S. sourced material)