Oxalic acid dihydrate, 99+%, extra pure, Thermo Scientific Chemicals
Oxalic acid dihydrate, 99+%, extra pure, Thermo Scientific Chemicals
Oxalic acid dihydrate, 99+%, extra pure, Thermo Scientific Chemicals
Oxalic acid dihydrate, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Oxalic acid dihydrate, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 6153-56-6 | C2H6O6 | 126.064 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
100 g
Packaging:
View all
Plastic drum
Catalog number ACR129600050
Price (MYR)
2,993.00
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
2,993.00
Add to cart
Chemical Identifiers
CAS63769-58-4
IUPAC Name(2S)-3-[4-(benzyloxy)phenyl]-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Molecular FormulaC21H25NO5
InChI KeyZAVSPTOJKOFMTA-LGWFVXIRNA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%