4-Nitrotoluene, 99%, Thermo Scientific Chemicals
4-Nitrotoluene, 99%, Thermo Scientific Chemicals
4-Nitrotoluene, 99%, Thermo Scientific Chemicals
4-Nitrotoluene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitrotoluene, 99%, Thermo Scientific Chemicals
CAS: 99-99-0 | C7H7NO2 | 137.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
Packaging:
Glass bottle
Catalog number ACR129051000
Price (MYR)
375.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
375.00
Add to cart
Specifications
Chemical Name or Material4-Nitrotoluene
Name Note99%
CAS99-99-0
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger