Nitroterephthalic acid, 99+%, Thermo Scientific Chemicals
Nitroterephthalic acid, 99+%, Thermo Scientific Chemicals
Nitroterephthalic acid, 99+%, Thermo Scientific Chemicals
Nitroterephthalic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nitroterephthalic acid, 99+%, Thermo Scientific Chemicals
CAS: 610-29-7 | C8H5NO6 | 211.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR129000250
Price (MYR)
1,013.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,013.00
Add to cart
Chemical Identifiers
CAS100-17-4
IUPAC Name1-methoxy-4-nitrobenzene
Molecular FormulaC7H7NO3
InChI KeyBNUHAJGCKIQFGE-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Melting point51°C to 53°C
GC>=99.0 %
Appearance (Color)Green to brown