4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%, Thermo Scientific Chemicals
4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%, Thermo Scientific Chemicals
4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%, Thermo Scientific Chemicals
4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%, Thermo Scientific Chemicals
4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%, C6H4NNa2O6P.6H2O, CAS Number-333338-18-4
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number ACR128860100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Nitrophenyl phosphate, disodium salt, hexahydrate
CAS333338-18-4
Melting Point>300.0°C
ColorWhite to Yellow
Infrared SpectrumAuthentic