2-Nitrophenylhydrazine, 97%, moistened with ca 30% water
2-Nitrophenylhydrazine, 97%, moistened with ca 30% water
2-Nitrophenylhydrazine, 97%, moistened with ca 30% water
2-Nitrophenylhydrazine, 97%, moistened with ca 30% water
Thermo Scientific Chemicals
2-Nitrophenylhydrazine, 97%, moistened with ca 30% water
CAS: 3034-19-3 | C6H7N3O2 | 153.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR12883010010 gGlass bottle
Catalog number ACR128830100
Price (MYR)
721.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
721.00
Add to cart
Chemical Identifiers
CAS127-08-2
IUPAC Namepotassium acetate
Molecular FormulaC2H3KO2
InChI KeySCVFZCLFOSHCOH-UHFFFAOYSA-M
SMILES[K+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO4>=99.0 %
Appearance (Form)Crystalline or granular powder
Appearance (Color)White