4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals
4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals
4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals
4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals
CAS: 830-03-5 | C8H7NO4 | 181.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
View all
Plastic bottle
Catalog number ACR128750250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1116-98-9
SpecificationsSpecification SheetSpecification Sheet
FormLiquid or viscous liquid
Assay (GC)≥97.5%
Appearance (Color)Clear colorless to white to yellow
Refractive Index1.4180-1.4220 @ 20?C