4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals
4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals
4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals
4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals
CAS: 830-03-5 | C8H7NO4 | 181.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
View all
Glass bottle
Catalog number ACR128750050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Nitrophenyl acetate
Name Note97%
CAS830-03-5
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger