2-Nitrophenol, 99%, Thermo Scientific Chemicals
2-Nitrophenol, 99%, Thermo Scientific Chemicals
2-Nitrophenol, 99%, Thermo Scientific Chemicals
2-Nitrophenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Nitrophenol, 99%, Thermo Scientific Chemicals
CAS: 88-75-5 | C6H5NO3 | 139.11 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR128745000500 gGlass bottle
Catalog number ACR128745000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10026-12-7
IUPAC Nameniobium(5+) pentachloride
Molecular FormulaCl5Nb
InChI KeyYHBDIEWMOMLKOO-UHFFFAOYSA-I
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Nb+5]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤0.1% (1000 ppm)
Particle Size-100 Mesh
FormPowder
Appearance (Color)Yellow
Indicator
RUO – Research Use Only