Nitrobenzene, 99%, Extra Pure, Thermo Scientific Chemicals
Nitrobenzene, 99%, Extra Pure, Thermo Scientific Chemicals
Nitrobenzene, 99%, Extra Pure, Thermo Scientific Chemicals
Nitrobenzene, 99%, Extra Pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nitrobenzene, 99%, Extra Pure, Thermo Scientific Chemicals
CAS: 98-95-3 | C6H5NO2 | 123.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass bottle
Catalog number ACR128420025
Price (MYR)
896.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
896.00
Add to cart
Chemical Identifiers
CAS12055-23-1
IUPAC Namehafnium(4+) dioxidandiide
Molecular FormulaHfO2
InChI KeyWIHZLLGSGQNAGK-UHFFFAOYSA-N
SMILES[O--].[O--].[Hf+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Impurity contentZr<1.5%
Total Metal Impurities excluding Zr≤1%
Particle Size-325 mesh