Nitrobenzene, 99%, Extra Pure, Thermo Scientific Chemicals
Nitrobenzene, 99%, Extra Pure, Thermo Scientific Chemicals
Nitrobenzene, 99%, Extra Pure, Thermo Scientific Chemicals
Nitrobenzene, 99%, Extra Pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nitrobenzene, 99%, Extra Pure, Thermo Scientific Chemicals
CAS: 98-95-3 | C6H5NO2 | 123.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number ACR128420010
Price (MYR)
620.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
620.00
Add to cart
Chemical Identifiers
CAS3483-12-3
IUPAC Name(2R,3R)-1,4-disulfanylbutane-2,3-diol
Molecular FormulaC4H10O2S2
InChI KeyVHJLVAABSRFDPM-IMJSIDKUSA-N
SMILESO[C@@H](CS)[C@@H](O)CS
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Absorbance (280 nm, 0.1M, 1 cm): ≤ 0.08
Impurity contentOxidized DTT: ≤ 0.3%
Protease activityPasses test
Assay (unspecified)(SH): ≥ 99.5%
DNasePasses test