3-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
3-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
3-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
3-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
CAS: 99-61-6 | C7H5NO3 | 151.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number ACR128415000
Price (MYR)
943.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
943.00
Add to cart
Chemical Identifiers
CAS75-30-9
IUPAC Name2-iodopropane
Molecular FormulaC3H7I
InChI KeyFMKOJHQHASLBPH-UHFFFAOYSA-N
SMILESCC(C)I
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.0 %
Appearance (Color)Clear colorless to light yellow
Refractive index1.4972 to 1.4992 (20°C, 589 nm)