3-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
3-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
3-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
3-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
CAS: 99-61-6 | C7H5NO3 | 151.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Catalog number ACR128411000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS67-63-0
IUPAC Namepropan-2-ol
Molecular FormulaC3H8O
InChI KeyKFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILESCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.2 %
Appearance (Form)Clear liquid
Color scale=<10 APHA
Non-volatile matter=<0.001 %
GC>=99.50 %