2-Nitrobenzaldehyde, 99+%, Thermo Scientific Chemicals
2-Nitrobenzaldehyde, 99+%, Thermo Scientific Chemicals
2-Nitrobenzaldehyde, 99+%, Thermo Scientific Chemicals
2-Nitrobenzaldehyde, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Nitrobenzaldehyde, 99+%, Thermo Scientific Chemicals
CAS: 552-89-6 | C7H5NO3 | 151.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number ACR128400250
Price (MYR)
618.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
618.00
Add to cart
Chemical Identifiers
CAS133073-73-1
IUPAC Name(2S)-2-[(4-{[(2,4-diaminopteridin-6-yl)methyl](methyl)amino}phenyl)formamido]pentanedioic acid
Molecular FormulaC20H22N8O5
InChI KeyFBOZXECLQNJBKD-UGPWUYPHNA-N
SMILESCN(CC1=CN=C2N=C(N)N=C(N)C2=N1)C1=CC=C(C=C1)C(=O)N[C@@H](CCC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Specific optical rotationon anhydrous substance
Appearance (Color)Yellow to orange
HPLC>=98.5 %
Specific optical rotation+19° to +24° (20°C, 589 nm) (c=1, 14 g/l Na2CO3)