4-Nitroaniline, 99%, Thermo Scientific Chemicals
4-Nitroaniline, 99%, Thermo Scientific Chemicals
4-Nitroaniline, 99%, Thermo Scientific Chemicals
4-Nitroaniline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitroaniline, 99%, Thermo Scientific Chemicals
CAS: 100-01-6 | C6H6N2O2 | 138.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number ACR128371000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Nitroaniline
CAS100-01-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breat
View more
Indicator
RUO – Research Use Only