Mucic Acid 98%, Thermo Scientific Chemicals
Mucic Acid 98%, Thermo Scientific Chemicals
Mucic Acid 98%, Thermo Scientific Chemicals
Mucic Acid 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Mucic Acid 98%, Thermo Scientific Chemicals
CAS: 526-99-8 | C6H10O8 | 210.138 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR128041000100 gPlastic bottle
Catalog number ACR128041000
Price (MYR)
601.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
601.00
Add to cart
Chemical Identifiers
CAS88-06-2
IUPAC Name2,4,6-trichlorophenol
Molecular FormulaC6H3Cl3O
InChI KeyLINPIYWFGCPVIE-UHFFFAOYSA-N
SMILESOC1=C(Cl)C=C(Cl)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White to cream to yellow to pale brown
FormCrystals or powder or crystalline powder or granules
Melting Point (clear melt)62.0-69.0°C