Methyl sulfide, 99+%, extra pure, Thermo Scientific Chemicals
Methyl sulfide, 99+%, extra pure, Thermo Scientific Chemicals
Methyl sulfide, 99+%, extra pure, Thermo Scientific Chemicals
Methyl sulfide, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl sulfide, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 75-18-3 | C2H6S | 62.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
10 mL
1 L
Packaging:
Glass bottle
Catalog number ACR127760025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS617-86-7
IUPAC Nametriethylsilyl
Molecular FormulaC6H15Si
InChI KeyQXTIBZLKQPJVII-UHFFFAOYSA-N
SMILESCC[Si](CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=98.5 %
Refractive index1.4100 to 1.4120 (20°C, 589 nm)