alpha-Methylstyrene, 99%, stabilized, Thermo Scientific Chemicals
alpha-Methylstyrene, 99%, stabilized, Thermo Scientific Chemicals
alpha-Methylstyrene, 99%, stabilized, Thermo Scientific Chemicals
alpha-Methylstyrene, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
alpha-Methylstyrene, 99%, stabilized, Thermo Scientific Chemicals
CAS: 98-83-9 | C9H10 | 118.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
Packaging:
Glass bottle
Catalog number ACR127711000
Price (MYR)
445.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
445.00
Add to cart
Chemical Identifiers
CAS75-31-0
IUPAC Namepropan-2-amine
Molecular FormulaC3H9N
InChI KeyJJWLVOIRVHMVIS-UHFFFAOYSA-N
SMILESCC(C)N
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.3725 to 1.3755 (20°C, 589 nm)
Water=<0.5 % (K.F.)
GC>=98.5 %
Appearance (Form)Clear liquid