alpha-Methylstyrene, 99%, stabilized, Thermo Scientific Chemicals
alpha-Methylstyrene, 99%, stabilized, Thermo Scientific Chemicals
alpha-Methylstyrene, 99%, stabilized, Thermo Scientific Chemicals
alpha-Methylstyrene, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
alpha-Methylstyrene, 99%, stabilized, Thermo Scientific Chemicals
CAS: 98-83-9 | C9H10 | 118.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
Packaging:
Glass bottle
Catalog number ACR127710010
Price (MYR)
580.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
580.00
Add to cart
Chemical Identifiers
CAS13520-61-1
IUPAC Namenickel(2+) hexakis(methane) diperchlorate
Molecular FormulaC6H24Cl2NiO8
InChI KeyGWPPTBKPEOUCMN-UHFFFAOYSA-L
SMILESC.C.C.C.C.C.[Ni++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Ni 15.3û17.0%