2-(Methylthio)aniline, 97%, Thermo Scientific Chemicals
2-(Methylthio)aniline, 97%, Thermo Scientific Chemicals
2-(Methylthio)aniline, 97%, Thermo Scientific Chemicals
2-(Methylthio)aniline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-(Methylthio)aniline, 97%, Thermo Scientific Chemicals
CAS: 2987-53-3 | C7H9NS | 139.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR127110250
Price (MYR)
1,216.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,216.00
Add to cart
Chemical Identifiers
CAS90076-65-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Infrared spectrumConforms
Titration after Ion exchange>=98.5 %
Appearance (Form)Powder or crystals