Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
CAS: 96-33-3 | C4H6O2 | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
10 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number ACR126195000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5470-18-8
IUPAC Name2-chloro-3-nitropyridine
Molecular FormulaC5H3ClN2O2
InChI KeyUUOLETYDNTVQDY-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CN=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Crystalline powder
Melting point100°C to 103°C
Infrared spectrumConforms
HPLC>=99.0 %