Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
CAS: 96-33-3 | C4H6O2 | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number ACR126190100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS872-50-4
IUPAC Name1-methylpyrrolidin-2-one
Molecular FormulaC5H9NO
InChI KeySECXISVLQFMRJM-UHFFFAOYSA-N
SMILESCN1CCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Free amine=<0.01 %
Water=<0.1 % (K.F.)
Appearance (Form)Clear liquid
GC>=99.4 %