Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
CAS: 96-33-3 | C4H6O2 | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
10 mL
500 mL
1 L
Packaging:
Glass bottle
Catalog number ACR126190025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5808-22-0
IUPAC Name4,5-dihydroxynaphthalene-2,7-disulfonate
Molecular FormulaC10H6O8S2
InChI KeyHLVXFWDLRHCZEI-UHFFFAOYSA-L
SMILESOC1=CC(=CC2=CC(=CC(O)=C12)S([O-])(=O)=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Solution TestP.T.
FormPowder
Appearance (Color)White to grey, beige or brown
CommentSensitivity to formaldehyde P.T.
CommentSensitivity to nitrate P.T.