Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
CAS: 96-33-3 | C4H6O2 | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
10 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number ACR126190010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS78-92-2
IUPAC Namebutan-2-ol
Molecular FormulaC4H10O
InChI KeyBTANRVKWQNVYAZ-UHFFFAOYNA-N
SMILESCCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
Water=<0.5 %
Color scale=<10 APHA
GC>=98.5 %