4-Methoxyphenol, 99%, Thermo Scientific Chemicals
4-Methoxyphenol, 99%, Thermo Scientific Chemicals
4-Methoxyphenol, 99%, Thermo Scientific Chemicals
4-Methoxyphenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Methoxyphenol, 99%, Thermo Scientific Chemicals
CAS: 150-76-5 | C7H8O2 | 124.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
100 g
500 g
1 kg
Catalog number ACR126000051
Price (MYR)
3,371.00
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (MYR)
3,371.00
Add to cart
Chemical Identifiers
CAS16375-88-5
IUPAC NameN-[4-(hydroxymethyl)phenyl]acetamide
Molecular FormulaC9H11NO2
InChI KeyXEYORFKUJZEQCH-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=C(CO)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow
Melting Point (clear melt)118.5-124.5?C
Assay (HPLC)≥96.0%