4-Methoxyphenol, 99%, Thermo Scientific Chemicals
4-Methoxyphenol, 99%, Thermo Scientific Chemicals
4-Methoxyphenol, 99%, Thermo Scientific Chemicals
4-Methoxyphenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Methoxyphenol, 99%, Thermo Scientific Chemicals
CAS: 150-76-5 | C7H8O2 | 124.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
500 g
5 kg
Catalog number ACR126000010
Price (MYR)
999.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
999.00
Add to cart
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Trace Metal0.002% max.