p-Methoxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
p-Methoxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
p-Methoxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
p-Methoxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Methoxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
CAS: 830-09-1 | C10H10O3 | 178.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR125841000
Price (MYR)
1,552.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,552.00
Add to cart
Chemical Identifiers
CAS14187-32-7
IUPAC Name2,5,8,15,18,21-hexaoxatricyclo[20.4.0.0⁹,¹⁴]hexacosa-1(26),9,11,13,22,24-hexaene
Molecular FormulaC20H24O6
InChI KeyYSSSPARMOAYJTE-UHFFFAOYSA-N
SMILESC1COC2=CC=CC=C2OCCOCCOC2=CC=CC=C2OCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Infrared spectrumConforms
Melting point160°C to 164°C
HPLC>=98.0 %
Appearance (Form)Powder