p-Methoxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
p-Methoxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
p-Methoxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
p-Methoxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Methoxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
CAS: 830-09-1 | C10H10O3 | 178.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR125840250
Price (MYR)
625.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
625.00
Add to cart
Chemical Identifiers
CAS2439-35-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless to yellow or pale brown
Assay (GC)>97.5%
Refractive Index1.4360-1.4400 @ 20?C
Solution Test(10% v/v in diethyl ether) : Clear
Solution Test(10% v/v in methanol) : Clear