4-Methoxybenzylamine, 98%, Thermo Scientific Chemicals
4-Methoxybenzylamine, 98%, Thermo Scientific Chemicals
4-Methoxybenzylamine, 98%, Thermo Scientific Chemicals
4-Methoxybenzylamine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Methoxybenzylamine, 98%, Thermo Scientific Chemicals
CAS: 2393-23-9 | C8H11NO | 137.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number ACR125780250
Price (MYR)
516.00
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
516.00
Add to cart
Chemical Identifiers
CAS127-08-2
IUPAC Namepotassium acetate
Molecular FormulaC2H3KO2
InChI KeySCVFZCLFOSHCOH-UHFFFAOYSA-M
SMILES[K+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble material0.005%
Impurity contentCa 0.005%
Assay (unspecified)99% C₂H₃KO₂ min
Heavy Metals(ICP-OES) 5ppm
Impurity contentCl 0.003%