Methanesulfonyl chloride, 99.5%, Thermo Scientific Chemicals
Methanesulfonyl chloride, 99.5%, Thermo Scientific Chemicals
Methanesulfonyl chloride, 99.5%, Thermo Scientific Chemicals
Methanesulfonyl chloride, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methanesulfonyl chloride, 99.5%, Thermo Scientific Chemicals
CAS: 124-63-0 | CH3ClO2S | 114.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
Packaging:
Glass bottle
Catalog number ACR125642500
Price (MYR)
594.00
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
594.00
Add to cart
Chemical Identifiers
CAS76-60-8
IUPAC Name3,3-bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular FormulaC21H14Br4O5S
InChI KeyFRPHFZCDPYBUAU-UHFFFAOYSA-N
SMILESCC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
UV spectrophotometry(lambda-max @ pH 5.4) : 530-600
FormPowder
Appearance (Color)Cream or yellow to brown
CommentSuitable for use as an indicator