Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
CAS: 141-79-7 | C6H10O | 98.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number ACR125600025
Price (MYR)
1,107.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,107.00
Add to cart
Chemical Identifiers
CAS35897-34-8
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl]amino}-5-carbamimidamidopentanoic acid hydrochloride
Molecular FormulaC11H23ClN4O4
InChI KeyHDELGKMVZYHPPB-RZUXQYDTNA-N
SMILESCl.CC(C)(C)OC(=O)N[C@@H](CCCNC(N)=N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (HPLC)≥97.5%
FormPowder
Assay (HPLC)≥97.5%