Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
CAS: 141-79-7 | C6H10O | 98.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number ACR125600010
Price (MYR)
655.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
655.00
Add to cart
Chemical Identifiers
CAS924868-81-5
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%