2-Mercaptothiazoline, 98%, Thermo Scientific Chemicals
2-Mercaptothiazoline, 98%, Thermo Scientific Chemicals
2-Mercaptothiazoline, 98%, Thermo Scientific Chemicals
2-Mercaptothiazoline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Mercaptothiazoline, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR125560050
Price (MYR)
335.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
335.00
Add to cart
Specifications
Chemical Name or Material2-Mercaptothiazoline
Name Note98%
CAS96-53-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning