Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
CAS: 68-11-1 | C2H4O2S | 92.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
10 mL
250 mL
2.5 L
Packaging:
Glass bottle
Catalog number ACR125430010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS104-94-9
IUPAC Name4-methoxyaniline
Molecular FormulaC7H9NO
InChI KeyBHAAPTBBJKJZER-UHFFFAOYSA-N
SMILESCOC1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to brown
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Crystalline powder or granules or flakes
Melting point56°C to 59°C